Skip to Content
Merck
Zinc trifluoromethanesulfonate 98%

Zinc trifluoromethanesulfonate

Zinc trifluoromethanesulfonate 98%
Synonym(s):
Trifluoromethanesulfonic acid zinc salt, Zinc triflate, Zn(OTf)2
Linear Formula:
(CF3SO3)2Zn
CAS Number:
Molecular Weight:
363.53
EC Number:

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing