Skip to Content
Merck
Zileuton Related Compound B pharmaceutical secondary standard, certified reference material

Zileuton Related Compound B

Zileuton Related Compound B pharmaceutical secondary standard, certified reference material
Synonym(s):
2-(Benzo[b]thien-2-oyl)benzo[b]thiophene, Benzo[b]thien-2-yl ketone, Bis(benzo[b]thien-2-yl)methanone
Empirical Formula (Hill Notation):
C17H10OS2
CAS Number:
Molecular Weight:
294.39

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade