![Z-Dehydro-Ala-OMe ≥98.0% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/275/954/28abe411-1102-4fbf-903b-047b2abd8667/640/28abe411-1102-4fbf-903b-047b2abd8667.png)
Z-Dehydro-Ala-OMe
![Z-Dehydro-Ala-OMe ≥98.0% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/275/954/28abe411-1102-4fbf-903b-047b2abd8667/640/28abe411-1102-4fbf-903b-047b2abd8667.png)
Synonym(s):
N-Cbz-Dehydroalanine methyl ester, Z-Dehydroalanine methyl ester
Linear Formula:
C6H5CH2O2CNHC(=CH2)CO2CH3
CAS Number:
Molecular Weight:
235.24
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing