![Venlafaxine hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/152/348/0c3a1d18-9fd7-4b4d-b33c-ec86f82b4b31/640/0c3a1d18-9fd7-4b4d-b33c-ec86f82b4b31.png)
Venlafaxine hydrochloride
![Venlafaxine hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/152/348/0c3a1d18-9fd7-4b4d-b33c-ec86f82b4b31/640/0c3a1d18-9fd7-4b4d-b33c-ec86f82b4b31.png)
Synonym(s):
(+/-)-1-[2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride, Effexor, Venlafaxine hydrochloride
Empirical Formula (Hill Notation):
C17H27NO2 · HCl
CAS Number:
Molecular Weight:
313.86
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing