Skip to Content
Merck
Venlafaxine-D6 hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Venlafaxine-D6 hydrochloride solution

Venlafaxine-D6 hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C17D6H21NO2 · HCl
CAS Number:
Molecular Weight:
319.90
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique