Valnemulin impurity E
Synonym(s):
Pleuromutilin, Drosophilin B, Mutilin 14-glycolate, NSC 121145
Empirical Formula (Hill Notation):
C22H34O5
CAS Number:
Molecular Weight:
378.50
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing