![Turbonuclease from Serratia marcescens recombinant, expressed in E. coli](/deepweb/assets/sigmaaldrich/product/images/279/686/79b39ae3-14df-4933-a710-4a21092dc59c/640/79b39ae3-14df-4933-a710-4a21092dc59c.jpg)
Turbonuclease from Serratia marcescens
![Turbonuclease from Serratia marcescens recombinant, expressed in E. coli](/deepweb/assets/sigmaaldrich/product/images/279/686/79b39ae3-14df-4933-a710-4a21092dc59c/640/79b39ae3-14df-4933-a710-4a21092dc59c.jpg)
Synonym(s):
Endonuclease from Serratia marcescens
CAS Number:
Products
Brand
Biological Source
Physical Form
Recombinant Host
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing