Tryptic Soy Agar - Ready-to-use Bottles & Tubes
Products
Product Category
Brand
Color
Physical Form
Agency
Capacity (mL)
Quality Segment
Closure Type
Application
Package Size
Sterility
Sterilization Method
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing