Skip to Content
Merck
Tris(trimethylsilyl)silane 97%

Tris(trimethylsilyl)silane

Tris(trimethylsilyl)silane 97%
Synonym(s):
1,1,1,3,3,3-Hexamethyl-2-trimethylsilyl-trisilane, TTMSS
Linear Formula:
[(CH3)3Si]3SiH
CAS Number:
Molecular Weight:
248.66

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing