Skip to Content
Merck
Tris(trimethylsilyl) phosphate ≥99.0%, anhydrous, battery grade

Tris(trimethylsilyl) phosphate

Tris(trimethylsilyl) phosphate ≥99.0%, anhydrous, battery grade
Synonym(s):
Phosphoric acid tris(trimethylsilyl ester)
Linear Formula:
[(CH3)3SiO]3P(O)
CAS Number:
Molecular Weight:
314.54

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-2 of 2