Tris(dibenzylideneacetone)dipalladium(0)
Synonym(s):
Pd2(dba)3
Linear Formula:
(C6H5CH=CHCOCH=CHC6H5)3Pd2
CAS Number:
Molecular Weight:
915.72
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing