Skip to Content
Merck
Tripalmitin NIST®SRM®595A

Tripalmitin

Tripalmitin NIST®SRM®595A
Synonym(s):
1,2,3-Propanetriol tris(hexadecanoate), Glyceryl tripalmitate
Linear Formula:
[CH3(CH2)14COOCH2]2CHOCO(CH2)14CH3
CAS Number:
Molecular Weight:
807.32

Products

Feature
Formula Weight
Melting Point (°C)
Format
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing