![Trazodone Related Compound D certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/406/120/200b8f65-a2a2-498c-be81-8a809c4a2259/640/200b8f65-a2a2-498c-be81-8a809c4a2259.jpg)
Trazodone Related Compound D
![Trazodone Related Compound D certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/406/120/200b8f65-a2a2-498c-be81-8a809c4a2259/640/200b8f65-a2a2-498c-be81-8a809c4a2259.jpg)
Synonym(s):
2-{3-[4-(3-Bromophenyl)piperazin-1-yl]propyl}-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one hydrochloride
Empirical Formula (Hill Notation):
C19H22BrN5O · HCl
CAS Number:
Molecular Weight:
452.78
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2