Skip to Content
Merck
trans-4-Aminocyclohexanol hydrochloride 97%

trans-4-Aminocyclohexanol hydrochloride

trans-4-Aminocyclohexanol hydrochloride 97%
Synonym(s):
trans-4-Hydroxycyclohexylamine hydrochloride
Linear Formula:
H2NC6H10OH · HCl
CAS Number:
Molecular Weight:
151.63

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing