Skip to Content
Merck
trans-1,2-Dichloroethylene Pharmaceutical Secondary Standard; Certified Reference Material

trans-1,2-Dichloroethylene

trans-1,2-Dichloroethylene Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
trans-1,2-Dichloroethene, trans-Acetylene dichloride, trans-1,2-Dichloroethylene
Linear Formula:
ClCH=CHCl
CAS Number:
Molecular Weight:
96.94

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique