Skip to Content
Merck
TOYOPEARL® AF-Red-650ML Bulk Media phase Procion Red HE-3B, 65 μm particle size, bottle of 1 L

TOYOPEARL® AF-Red-650ML Bulk Media

TOYOPEARL® AF-Red-650ML Bulk Media phase Procion Red HE-3B, 65 μm particle size, bottle of 1 L
Synonym(s):
TOYOPEARL® AF-Red-650M Bulk Media

Products

Manufacturer
pH Value
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1