
Toltrazuril

Synonym(s):
1-Methyl-3-{3-methyl-4-[4-(trifluoromethylthio)phenoxy]phenyl}-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, Baycox
Empirical Formula (Hill Notation):
C18H14F3N3O4S
CAS Number:
Molecular Weight:
425.38
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing