Skip to Content
Merck
Tolclofos-methyl PESTANAL®, analytical standard

Tolclofos-methyl

Tolclofos-methyl PESTANAL®, analytical standard
Synonym(s):
O-(2,6-Dichloro-4-methylphenyl) O,O-dimethyl phosphorothioate
Empirical Formula (Hill Notation):
C9H11Cl2O3PS
CAS Number:
Molecular Weight:
301.13
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing