Skip to Content
Merck
Titanium(IV) ethoxide technical grade

Titanium(IV) ethoxide

Titanium(IV) ethoxide technical grade
Synonym(s):
Tetraethyl orthotitanate, Tetraethyl titanate
Linear Formula:
Ti(OC2H5)4
CAS Number:
Molecular Weight:
228.11

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing