Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tin-silver alloy, Sn96Ag4
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
GF74592692
Tin-silver alloy, Sn96Ag4,
1m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF26045412
Tin-silver alloy, Sn96Ag4,
200m wire, 0.27mm diameter, as drawn
Expand
View Pricing
GF86659521
Tin-silver alloy, Sn96Ag4,
50m wire, 0.27mm diameter, as drawn
Expand
View Pricing
GF90813900
Tin-silver alloy, Sn96Ag4,
2m wire, 1.5mm diameter, as drawn
Expand
View Pricing
GF73503299
Tin-silver alloy, Sn96Ag4,
20m wire, 0.50mm diameter, as drawn
Expand
View Pricing
GF96742008
Tin-silver alloy, Sn96Ag4,
10m wire, 0.50mm diameter, as drawn
Expand
View Pricing
GF90106141
Tin-silver alloy, Sn96Ag4,
5m wire, 0.50mm diameter, as drawn
Expand
View Pricing
GF69667060
Tin-silver alloy, Sn96Ag4,
5m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF70679335
Tin-silver alloy, Sn96Ag4,
10m wire, 1.5mm diameter, as drawn
Expand
View Pricing
GF88799933
Tin-silver alloy, Sn96Ag4,
1m wire, 1.5mm diameter, as drawn
Expand
View Pricing
GF30786127
Tin-silver alloy, Sn96Ag4,
50m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF33672102
Tin-silver alloy, Sn96Ag4,
20m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF25175280
Tin-silver alloy, Sn96Ag4,
5m wire, 1.5mm diameter, as drawn
Expand
View Pricing
GF25651740
Tin-silver alloy, Sn96Ag4,
10m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF79253400
Tin-silver alloy, Sn96Ag4,
50m wire, 0.50mm diameter, as drawn
Expand
View Pricing
GF98626472
Tin-silver alloy, Sn96Ag4,
20m wire, 0.27mm diameter, as drawn
Expand
View Pricing
GF80175449
Tin-silver alloy, Sn96Ag4,
20m wire, 1.5mm diameter, as drawn
Expand
View Pricing
GF82917658
Tin-silver alloy, Sn96Ag4,
100m wire, 0.50mm diameter, as drawn
Expand
View Pricing
GF76615252
Tin-silver alloy, Sn96Ag4,
100m wire, 0.27mm diameter, as drawn
Expand
View Pricing
GF00998309
Tin-silver alloy, Sn96Ag4,
10m wire, 0.27mm diameter, as drawn
Expand
View Pricing