
Tin(IV) chloride solution

Synonym(s):
Stannic chloride, Tin tetrachloride
Linear Formula:
SnCl4
CAS Number:
Molecular Weight:
260.52
Products
Product Category
Formula Weight
Physical Form
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing