Skip to Content
Merck
Tin(IV) chloride pentahydrate 98%

Tin(IV) chloride pentahydrate

Tin(IV) chloride pentahydrate 98%
Synonym(s):
Tin tetrachloride
Linear Formula:
SnCl4 · 5H2O
CAS Number:
Molecular Weight:
350.60

Products

Product Category
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing