Skip to Content
Merck
vibrant-m

Tinidazole impurity B

vibrant-m
Synonym(s):
1-[2-(Ethylsulfonyl)ethyl]-2-methyl-4-nitro-1H-imidazole
Empirical Formula (Hill Notation):
C8H13N3O4S
CAS Number:
Molecular Weight:
247.27

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing