Thifensulfuron-methyl
Synonym(s):
Harmony
Empirical Formula (Hill Notation):
C12H13N5O6S2
CAS Number:
Molecular Weight:
387.39
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2