Skip to Content
Merck
Tetrabutylammonium difluorotriphenylsilicate 97%

Tetrabutylammonium difluorotriphenylsilicate

Tetrabutylammonium difluorotriphenylsilicate 97%
Synonym(s):
TBAT
Linear Formula:
(CH3CH2CH2CH2)4N[(C6H5)3SiF2]
CAS Number:
Molecular Weight:
539.86

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing