Skip to Content
Merck
tert-Butyl(chloro)diphenylsilane 98%

tert-Butyl(chloro)diphenylsilane

tert-Butyl(chloro)diphenylsilane 98%
Synonym(s):
tert-Butyldiphenylchlorosilane, TBDPSCl
Linear Formula:
(CH3)3CSi(C6H5)2Cl
CAS Number:
Molecular Weight:
274.86
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing