Skip to Content
Merck
tert-Butanol-d10 reagent grade, ≥99 atom % D, ≥99% (CP)

tert-Butanol-d10

tert-Butanol-d10 reagent grade, ≥99 atom % D, ≥99% (CP)
Synonym(s):
2-Methyl-2-propanol-d10, tert-Butyl alcohol-d10
Linear Formula:
(CD3)3COD
CAS Number:
Molecular Weight:
84.18
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing