Tebupirimfos
Empirical Formula (Hill Notation):
C13H23N2O3PS
CAS Number:
Molecular Weight:
318.37
Products
Product Category
Brand
Formula Weight
Product Line
Format
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing