![Tebuconazole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/355/753/04753f6b-e246-4ef8-97ba-4a3be71a500e/640/04753f6b-e246-4ef8-97ba-4a3be71a500e.png)
Tebuconazole
![Tebuconazole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/355/753/04753f6b-e246-4ef8-97ba-4a3be71a500e/640/04753f6b-e246-4ef8-97ba-4a3be71a500e.png)
Synonym(s):
1-(4-Chlorophenyl)-4,4-dimethyl-3-(1H-1,2,4-triazol-1-ylmethyl)-3-pentanol
Empirical Formula (Hill Notation):
C16H22ClN3O
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2