Skip to Content
Merck
Sulfur trioxide triethylamine complex technical, ≥95% sulfur basis

Sulfur trioxide triethylamine complex

Sulfur trioxide triethylamine complex technical, ≥95% sulfur basis
Synonym(s):
NSC 68185, NSC 8168, Triethylamine sulfur trioxide complex
Linear Formula:
(CH3CH2)3N · SO3
CAS Number:
Molecular Weight:
181.25

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing