![Sulfinpyrazone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/439/503/33405a69-bd11-449c-94ca-187747a536f0/640/33405a69-bd11-449c-94ca-187747a536f0.png)
Sulfinpyrazone
![Sulfinpyrazone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/439/503/33405a69-bd11-449c-94ca-187747a536f0/640/33405a69-bd11-449c-94ca-187747a536f0.png)
Synonym(s):
(±)-Sulfinpyrazone, 1,2-Diphenyl-4-(phenylsulfinylethyl)-3,5-pyrazolidinedione, Diphenylpyrazone
Empirical Formula (Hill Notation):
C23H20N2O3S
CAS Number:
Molecular Weight:
404.48
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing