Sucrose octaacetate
Synonym(s):
D-(+)-Sucrose octaacetate, Sucrose octaacetate
Empirical Formula (Hill Notation):
C28H38O19
CAS Number:
Molecular Weight:
678.59
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing