Sucrose acetate isobutyrate solution
Empirical Formula (Hill Notation):
C40H62O19
CAS Number:
Molecular Weight:
846.91
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing