Skip to Content
Merck
Streptozocin ≥75% α-anomer basis, ≥98% (HPLC), powder

Streptozocin

Streptozocin ≥75% α-anomer basis, ≥98% (HPLC), powder
Synonym(s):
N-(Methylnitrosocarbamoyl)-α-D-glucosamine, Streptozotocin
Empirical Formula (Hill Notation):
C8H15N3O7
CAS Number:
Molecular Weight:
265.22
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing