![Sotalol Related Compound B certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/208/211/70431e77-7a5b-4e6b-af80-d94184b4302f/640/70431e77-7a5b-4e6b-af80-d94184b4302f.jpg)
Sotalol Related Compound B
![Sotalol Related Compound B certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/208/211/70431e77-7a5b-4e6b-af80-d94184b4302f/640/70431e77-7a5b-4e6b-af80-d94184b4302f.jpg)
Synonym(s):
N-(4-Formylphenyl)methanesulfonamide
Empirical Formula (Hill Notation):
C8H9NO3S
CAS Number:
Molecular Weight:
199.23
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2