Magnesium sulfate solution
Empirical Formula (Hill Notation):
MgSO4
CAS Number:
Molecular Weight:
120.37
Products
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Sterilization Method
Showing 1-2 of 2
Compare
Product Number
Description
Pricing