Skip to Content
Merck
Sodium phosphate monobasic monohydrate puriss. p.a., ACS reagent, ≥99.0% (T)

Sodium phosphate monobasic monohydrate

Sodium phosphate monobasic monohydrate puriss. p.a., ACS reagent, ≥99.0% (T)
Synonym(s):
Monosodium phosphate, Sodium dihydrogen phosphate monohydrate
Linear Formula:
NaH2PO4 · H2O
CAS Number:
Molecular Weight:
137.99
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Technique