Skip to Content
Merck
Sodium ionophore X Selectophore™, function tested

Sodium ionophore X

Sodium ionophore X Selectophore™, function tested
Synonym(s):
4-tert-Butylcalix[4]arene-tetraacetic acid tetraethyl ester
Empirical Formula (Hill Notation):
C60H80O12
CAS Number:
Molecular Weight:
993.27

Products

Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing