![Sodium dodecylbenzenesulfonate Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/116/495/4d5ee0b4-aa79-4133-9856-44a7c6a6837d/640/4d5ee0b4-aa79-4133-9856-44a7c6a6837d.png)
Sodium dodecylbenzenesulfonate
![Sodium dodecylbenzenesulfonate Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/116/495/4d5ee0b4-aa79-4133-9856-44a7c6a6837d/640/4d5ee0b4-aa79-4133-9856-44a7c6a6837d.png)
Synonym(s):
Dodecylbenzenesulfonic acid sodium salt
Linear Formula:
CH3(CH2)11C6H4SO3Na
Products
Product Category
Brand
Formula Weight
Physical Form
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing