Skip to Content
Merck
Sodium chloride Redi-Dri™, ≥99%, DNase, RNase, and protease (none detected), free-flowing

Sodium chloride

Sodium chloride Redi-Dri™, ≥99%, DNase, RNase, and protease (none detected), free-flowing
Synonym(s):
Halite, NaCl, Sodium chloride
Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Research Area
Quality Segment
Application
Special Grade
Technique
Showing 1-20 of 33
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2