
Sodium bisulfate monohydrate

Synonym(s):
Sodium hydrogen sulfate monohydrate
Linear Formula:
NaHSO4 · H2O
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing