Skip to Content
Merck
Sodium 1-hexanesulfonate solution suitable for ion pair chromatography, concentrate, LiChropur™, ampule

Sodium 1-hexanesulfonate solution

Sodium 1-hexanesulfonate solution suitable for ion pair chromatography, concentrate, LiChropur™, ampule
Synonym(s):
1-Hexanesulfonic acid sodium salt
Linear Formula:
CH3(CH2)4CH2SO3Na
CAS Number:
Molecular Weight:
188.22

Products

Brand
Formula Weight
Quality Segment
Package Size
Technique