Silicon dioxide, alumina doped
Synonym(s):
silicic anhydride, alumina doped preparation
Linear Formula:
(SiO2)x(Al2O3)y
Molecular Weight:
60.08
Products
Brand
Formula Weight
pH Value
Physical Form
Purity
Chemical Composition
Showing 1-1 of 1
Compare
Product Number
Description
Pricing