
(S)-GLYCIDYL PHTHALIMIDE

Synonym(s):
(S)-(+)-N-(2,3-Epoxypropyl)phthalimide, (S)-N-Glycidylphthalimide
Empirical Formula (Hill Notation):
C11H9NO3
CAS Number:
Molecular Weight:
203.19
Products
Formula Weight
Manufacturer
Agency
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing