Skip to Content
Merck
Sevoflurane Related Compound C certified reference material, pharmaceutical secondary standard

Sevoflurane Related Compound C

Sevoflurane Related Compound C certified reference material, pharmaceutical secondary standard
Synonym(s):
1,1,1,3,3,3-Hexafluoro-2-propanol, HFP, Hexafluoroisopropanol
Linear Formula:
(CF3)2CHOH
CAS Number:
Molecular Weight:
168.04

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade