Skip to Content
Merck
Salinomycin from Streptomyces albus, ≥98% (HPLC)

Salinomycin

Salinomycin from Streptomyces albus, ≥98% (HPLC)
Empirical Formula (Hill Notation):
C42H70O11
CAS Number:
Molecular Weight:
751.00
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing