![Salbutamol sulfate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/304/728/2b6cca54-a064-4e07-90ea-af29137294f6/640/2b6cca54-a064-4e07-90ea-af29137294f6.png)
Salbutamol sulfate
![Salbutamol sulfate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/304/728/2b6cca54-a064-4e07-90ea-af29137294f6/640/2b6cca54-a064-4e07-90ea-af29137294f6.png)
Synonym(s):
Salbutamol hemisulfate salt, α1-[(tert-Butylamino)methyl]-4-hydroxy-1,3-benzenedimethanol hemisulfate salt, Albuterol sulfate
Empirical Formula (Hill Notation):
C13H21NO3 · 0.5H2SO4
CAS Number:
Molecular Weight:
288.35
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing