(S)-(−)-6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride
Empirical Formula (Hill Notation):
C12H15NO4 · HCl
CAS Number:
Molecular Weight:
273.71
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing