![Ruscogenin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/292/833/b543b1d1-6c82-4926-8f3e-0b2d3ef2757b/640/b543b1d1-6c82-4926-8f3e-0b2d3ef2757b.png)
Ruscogenin
![Ruscogenin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/292/833/b543b1d1-6c82-4926-8f3e-0b2d3ef2757b/640/b543b1d1-6c82-4926-8f3e-0b2d3ef2757b.png)
Synonym(s):
(1β,3β,25R)-Spirost-5-ene-1,3-diol
Empirical Formula (Hill Notation):
C27H42O4
CAS Number:
Molecular Weight:
430.62
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing