Skip to Content
Merck
RuCl(p-cymene)[(R,R)-Ts-DPEN]

RuCl(p-cymene)[(R,R)-Ts-DPEN]

RuCl(p-cymene)[(R,R)-Ts-DPEN]
Synonym(s):
[N-[(1R,2R)-2-(Amino-κN)-1,2-diphenylethyl]-4-methylbenzenesulfonamidato-κN]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]-ruthenium
Empirical Formula (Hill Notation):
C31H35ClN2O2RuS
CAS Number:
Molecular Weight:
636.21

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing